Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks Wide Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07025046
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10305841
  • 122.96€ / Pack of 10
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • With graduation
  • Material: DURAN™
Specifications
Specifications
50 mm
Beaded rim and graduation
79 mm
200 mL
10/Pk.