missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7044868
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
533-87-9
C16H32O5
304.427
MFCD00135596
MEHUJCGAYMDLEL-LSDHHAIUSA-N
aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
7269316
(9S,10R)-9,10,16-trihydroxyhexadecanoic acid
C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O
This item is not returnable. View return policy
533-87-9
C16H32O5
304.427
MFCD00135596
MEHUJCGAYMDLEL-LSDHHAIUSA-N
aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
7269316
(9S,10R)-9,10,16-trihydroxyhexadecanoic acid
C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O

α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

Solubility
Soluble in water, alcohol, acetic acid and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 97°C to 101°C
Odor Odorless
Quantity 5 g
Beilstein 1727724
Merck Index 14,231
Solubility Information Soluble in water,alcohol,acetic acid and acetone.
Formula Weight 304.43
Percent Purity 95%
Chemical Name or Material (+/-)-erythro-Aleuritic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'