missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ES Type 14 Tag Out

Brand:  Brady™ 284433

Additional Details : Weight : 0.00100kg

Product Code. 11756885

  • 35.95€ / 1 pack
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

ES Type 14 Tag Out
Polyester
5.71 in.
10 Pk.
-40°C to +100°C
3.35 in.
Black/Yellow on White
Safety Identification
145 mm
-40°F to +212°F
0.249 mm
85 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions