Learn More
Notrax ESD Common Grounding Cord
Product Code. p-7227334
Click to view available options
Connector Types:
Female Connector
Ring Connector
Unit Size:
Each
Product Code. 17711918
Brand: Notrax 052SCG0100
Only null left
Add to basket
Add to basket
Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment.
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
  • 45 m cable connects wrist bands or bench mat to ground
  • Assures that all parts are at the same ground potential
  • Easy to install, simply snap it to a mat, plug in the wrist strap coil cord and connect terminal to ground
  • Resistor 1 Meg mat to ground
  • Available with a ring terminal or 10 mm female socket
Specifications
Color Black
Product Type Grounding Cord
Material Wire
Connector Types Ring Connector