missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC), Honeywell™

Brand:  Honeywell Research Chemicals 24194-5L

Additional Details : CAS : 64-17-5 Poids : 3.94500kg Transport : UN number : 1170 Chem class : 3 Pack group : II

Quantity 5L

 View more versions of this product

Product Code. 15652540

  • 108.13€ / 5 L


En stock, prêt à être expédié : 68
Add to basket

Specifications

Chemical Identifiers

64-17-5
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
C2H6O
MFCD00003568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO

Specifications

Ethanol
78-93-3
<5.0000%
CH3CH2OH
UN1170
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46
0.789 g/mL at 25 °C(lit.)
683°F
44.6mmHg (20°C)
12°C
-114°C (literature)
Absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
≥1.0000%
C2H6O
MFCD00003568
1718733
Completely souble in water
CCO
46.069
CHEBI:16236
>98%
19% (60°F)
Poly Bottle
1.59 (vs air)
78°C to 79°C
5L
Videos
Documents