Learn More
Alla France™ Ethanol Hydrometer
Click to view available options
Brix Range:
20 to 30%
60 to 70%
90 to 100%
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Type | Ethanol Hydrometer |
| Accuracy | ±0.1% |
| Length (Metric) | 350 mm |
| Brix Range | 60 to 70% |