Learn More
Memmert™ Ethernet Connection Cable for Computer Interface for UN30
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
Description
- 5 m
- Accessory
Specifications
Specifications
| For Use With (Equipment) | UN30 Oven |
| Type | USB Connection Cable |
| Length (Metric) | 5 m |