missing translation for 'onlineSavingsMsg'
Learn More

Ethinylestradiol, 98%, Thermo Scientific Chemicals

Catalog Number p-7077892
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
57-63-6
C20H24O2
296.41
MFCD00003690
BFPYWIDHMRZLRN-SLHNCBLASA-N
ethinyl estradiol, ethynylestradiol, ethynyl estradiol, ethinylestradiol, ethinyloestradiol, ginestrene, 17-ethinylestradiol, progynon c, ethinoral, eticyclin
5991
CHEBI:4903
(8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C
This item is not returnable. View return policy
57-63-6
C20H24O2
296.41
MFCD00003690
BFPYWIDHMRZLRN-SLHNCBLASA-N
ethinyl estradiol, ethynylestradiol, ethynyl estradiol, ethinylestradiol, ethinyloestradiol, ginestrene, 17-ethinylestradiol, progynon c, ethinoral, eticyclin
5991
CHEBI:4903
(8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 180.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 296.41
Percent Purity 98%
Chemical Name or Material Ethinylestradiol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'