Learn More
Alfa Aesar™ Ethyl 2-amino-4-(4-chlorophenyl)-5-methylthiophene-3-carboxylate, 96%
Brand: Alfa Aesar™ H33651.03
Additional Details : CAS : 350989-77-4 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
350989-77-4 | |
295.781 | |
PJCFNTKRSCPMFG-UHFFFAOYSA-N | |
711209 | |
CCOC(=O)C1=C(SC(=C1C2=CC=C(C=C2)Cl)C)N |
C14H14ClNO2S | |
MFCD01921958 | |
ethyl 2-amino-4-4-chlorophenyl-5-methylthiophene-3-carboxylate, ethyl 2-amino-4-4-chlorophenyl-5-methyl-3-thiophenecarboxylate, 3-thiophenecarboxylicacid, 2-amino-4-4-chlorophenyl-5-methyl-, ethyl ester, cbmicro_046608 | |
ethyl 2-amino-4-(4-chlorophenyl)-5-methylthiophene-3-carboxylate |
Specifications
Ethyl 2-amino-4-(4-chlorophenyl)-5-methylthiophene-3-carboxylate | |
1g | |
C14H14ClNO2S | |
ethyl 2-amino-4-4-chlorophenyl-5-methylthiophene-3-carboxylate, ethyl 2-amino-4-4-chlorophenyl-5-methyl-3-thiophenecarboxylate, 3-thiophenecarboxylicacid, 2-amino-4-4-chlorophenyl-5-methyl-, ethyl ester, cbmicro_046608 | |
CCOC(=O)C1=C(SC(=C1C2=CC=C(C=C2)Cl)C)N | |
295.781 | |
295.79 |
92°C to 95°C | |
350989-77-4 | |
MFCD01921958 | |
PJCFNTKRSCPMFG-UHFFFAOYSA-N | |
ethyl 2-amino-4-(4-chlorophenyl)-5-methylthiophene-3-carboxylate | |
711209 | |
96% |