Learn More
Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%, Alfa Aesar™
Brand: Alfa Aesar H34197.06
Additional Details : CAS : 4506-71-2 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
4506-71-2 | |
225.306 | |
CDYVTVLXEWMCHU-UHFFFAOYSA-N | |
78262 | |
CCOC(=O)C1=C(SC2=C1CCCC2)N |
C11H15NO2S | |
MFCD00005862 | |
ethyl 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid ethyl ester, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid ethyl ester, benzo b thiophene-3-carboxylic acid, 2-amino-4,5,6,7-tetrahydro-, ethyl ester, ethyl2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, methylaminotetrahydrobenzothiophenecarboxylate, 2-amino-3-carbethoxy-4,5-tetramethylenethiophene, ethyl 2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxylate, ethyl-2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate | |
ethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate |
Specifications
Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate | |
5g | |
C11H15NO2S | |
ethyl 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid ethyl ester, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid ethyl ester, benzo b thiophene-3-carboxylic acid, 2-amino-4,5,6,7-tetrahydro-, ethyl ester, ethyl2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, methylaminotetrahydrobenzothiophenecarboxylate, 2-amino-3-carbethoxy-4,5-tetramethylenethiophene, ethyl 2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxylate, ethyl-2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate | |
CCOC(=O)C1=C(SC2=C1CCCC2)N | |
225.306 | |
225.31 |
114°C to 116°C | |
4506-71-2 | |
MFCD00005862 | |
CDYVTVLXEWMCHU-UHFFFAOYSA-N | |
ethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate | |
78262 | |
98% |