missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoro-2-butynoate, 97%, Thermo Scientific Chemicals

Catalog Number 15410228
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
79424-03-6
C6H5F3O2
166.099
MFCD00192167
SFDRHPQGYUYYNX-UHFFFAOYSA-N
3597254
ethyl 4,4,4-trifluorobut-2-ynoate
CCOC(=O)C#CC(F)(F)F
This item is not returnable. View return policy
79424-03-6
C6H5F3O2
166.099
MFCD00192167
SFDRHPQGYUYYNX-UHFFFAOYSA-N
3597254
ethyl 4,4,4-trifluorobut-2-ynoate
CCOC(=O)C#CC(F)(F)F

Ethyl 4,4,4-trifluoro-2-butynoate is the suitable reagent used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. It may be used in the preparation of ethyl (Z)-3-iodo-4,4,4-trifluoro-2-butenoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4,4,4-trifluoro-2-butynoate is the suitable reagent used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. It may be used in the preparation of ethyl (Z)-3-iodo-4,4,4-trifluoro-2-butenoate.

Solubility
Soluble in water (partly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.162
Boiling Point 96°C to 98°C
Flash Point 6°C (43°F)
Linear Formula CF3C≡CCO2CH2CH3
Refractive Index 1.35
Quantity 1 g
UN Number UN3272
Beilstein 3539414
Synonym ethyl 4,4,4-trifluoro-2-butynoate, 4,4,4-trifluoro-2-butynoic acid ethyl ester, 4,4,4-trifluoro-but-2-ynoic acid ethyl ester, pubchem10880, acmc-1bc5r, ethyl4,4,4-trifluoro-2-butynoate, ethyl 4,4,4-trifluoro-2-butynate, trifluoro-2-butynoic acid ethyl ester, ethyl 4,4,4-trifluorobutynoate, ethyl 4,4,4-tris fluoranyl but-2-ynoate
Solubility Information Soluble in water (partly).
Formula Weight 166.1
Percent Purity 97%
Chemical Name or Material Ethyl 4,4,4-trifluoro-2-butynoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'