missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-6-hydroxy-5-nitronicotinate, 97%, Thermo Scientific™

Catalog Number p-7052432
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1210835-74-7
C8H7ClN2O5
246.603
MFCD22989339
FETZVQJUBMRFJE-UHFFFAOYSA-N
ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate, ethyl 4-chloro-6-hydroxy-5-nitronicotinate, ethyl 4-chloro-6-hydroxy-5-nitropyridine-3-carboxylate, ethylchloronitrooxodihydropyridinecarboxylate, 4-chloro-6-hydroxy-5-nitronicotinic acid ethyl ester, ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 4-chloro-5-nitro-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid ethyl ester
59225818
ethyl 4-chloro-5-nitro-6-oxo-1H-pyridine-3-carboxylate
CCOC(=O)C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]
This item is not returnable. View return policy
1210835-74-7
C8H7ClN2O5
246.603
MFCD22989339
FETZVQJUBMRFJE-UHFFFAOYSA-N
ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate, ethyl 4-chloro-6-hydroxy-5-nitronicotinate, ethyl 4-chloro-6-hydroxy-5-nitropyridine-3-carboxylate, ethylchloronitrooxodihydropyridinecarboxylate, 4-chloro-6-hydroxy-5-nitronicotinic acid ethyl ester, ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 4-chloro-5-nitro-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid ethyl ester
59225818
ethyl 4-chloro-5-nitro-6-oxo-1H-pyridine-3-carboxylate
CCOC(=O)C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 176°C to 178°C
Color Pink to White
Quantity 250 mg
Formula Weight 246.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 4-chloro-6-hydroxy-5-nitronicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'