missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Reagent Grade, ≥99%, Honeywell™

Catalog Number p-7120420
Click to view available options
Quantity:
5 L
25 L
Packaging:
Poly Bottle
Stainless Steel Can
This item is not returnable. View return policy
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
This item is not returnable. View return policy
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -84°C
Color Colorless
Linear Formula CH3COOC2H5
UN Number UN1173
Beilstein 506104
Formula Weight 88.11g/mol
Percent Purity ≥99%
Density 0.902g/mL at 25°C
Boiling Point 77.5°C
Quantity 5 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.373
Vapor Pressure 73mmHg (20°C)
Chemical Name or Material Ethyl acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'