Learn More
Ethyl cyanoformate, 99%
Description
Ethyl cyanoformate is used as reagent in the preparation of N-substituted amindinoformic acid and ethyl-4-quinazoline -2-carboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl cyanoformate is used as reagent in the preparation of N-substituted amindinoformic acid and ethyl-4-quinazoline -2-carboxylate.
Solubility
Miscible with chloroform and methanol.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Density | 1.003 |
| Boiling Point | 115°C to 116°C |
| Flash Point | 24°C (75°F) |
| Refractive Index | 1.382 |
| Quantity | 100 g |
| UN Number | UN3275 |
| Beilstein | 635955 |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Solubility Information | Miscible with chloroform and methanol. |
| Formula Weight | 99.09 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.