missing translation for 'onlineSavingsMsg'
Learn More

Ethyl imidazo[1,2-a]pyridine-6-carboxylate, 95%, Alfa Aesar™

Brand:  Alfa Aesar H35696.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15409045

  • 36.37€ / 250mg


Estimated Shipment
22-04-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

158001-04-8
190.202
QPESMSWFRPZADJ-UHFFFAOYSA-N
11830209
CCOC(=O)C1=CN2C=CN=C2C=C1
C10H10N2O2
MFCD13176746
ethyl imidazo 1,2-a pyridine-6-carboxylate, imidazo 1,2-a pyridine-6-carboxylic acid ethyl ester, imidazo 1,2-a pyridine-6-carboxylic acid, ethyl ester, 6-carboethoxyimidazo 1,2-a pyridine, 6-carboethoxy imidazo 1,2-a-pyridine
ethyl imidazo[1,2-a]pyridine-6-carboxylate

Specifications

Ethyl imidazo[1,2-a]pyridine-6-carboxylate
250mg
C10H10N2O2
ethyl imidazo 1,2-a pyridine-6-carboxylate, imidazo 1,2-a pyridine-6-carboxylic acid ethyl ester, imidazo 1,2-a pyridine-6-carboxylic acid, ethyl ester, 6-carboethoxyimidazo 1,2-a pyridine, 6-carboethoxy imidazo 1,2-a-pyridine
CCOC(=O)C1=CN2C=CN=C2C=C1
190.202
190.2
97°C to 99°C
158001-04-8
MFCD13176746
QPESMSWFRPZADJ-UHFFFAOYSA-N
ethyl imidazo[1,2-a]pyridine-6-carboxylate
11830209
95%
Videos
SDS Documents