missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Indole-3-carboxylate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Ethyl Indole-3-carboxylate
Specifications
Specifications
| Chemical Name or Material | Ethyl indole-3-carboxylate |
| CAS | 776-41-0 |
| Analyte or Component Names | Ethyl indole-3-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H11 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl Indole-3-carboxylate, 1H-Indole-3-carboxylic acid ethyl ester, Indole-3-carboxylic acid ethyl ester, 3-(Ethoxycarbonyl)indole, Ethyl 1H-indole-3-carboxylate, Ethyl indole-3-carboxylate, NSC 63796 |
| InChI Formula | InChI=1S/C11H11NO2/c1-2-14-11(13)9-7-12-10-6-4-3-5-8(9)10/h3-7,12H,2H2,1H3 |
| SMILES | CCOC(=O)c1c[nH]c2ccccc12 |
| IUPAC Name | ethyl 1H-indole-3-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction