missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isocyanatoacetate, TRC

Product Code. 30383475
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30383475

Brand: Toronto Research Chemicals TRCE678885500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Isocyanatoacetate
CAS 2949-22-6
Molecular Formula C5H7NO3
Quantity 500 mg
Synonym Glycine ethyl ester isocyanate, Isocyanatoacetic acid ethyl ester, NSC 56429, NSC 6276, Carbethoxymethyl isocyanate, Ethoxycarbonylmethyl isocyanate
InChI Formula InChI=1S/C5H7NO3/c1-2-9-5(8)3-6-4-7/h2-3H2,1H3
SMILES CCOC(=O)CN=C=O
IUPAC Name ethyl 2-isocyanatoacetate
Molecular Weight (g/mol) 129.11
Formula Weight 129.0426
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.