missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalaninate (Nateglinide Ethyl Ester), Mikromol™
Description
Ethyl N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalaninate (Nateglinide Ethyl Ester)
Specifications
Specifications
| Chemical Name or Material | Ethyl N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalaninate (Nateglinide Ethyl Ester) |
| CAS | 187728-85-4 |
| Analyte or Component Names | Ethyl N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalaninate (Nateglinide Ethyl Ester) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H31 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl N-[[trans-4-(Propan-2-yl)cyclohexyl]carbonyl]-D-phenylalaninate, Nateglinide Imp. G (EP), Nateglinide Ethyl Ester, Ethyl N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalaninate, Nateglinide Impurity G |
| InChI Formula | InChI=1S/C21H31NO3/c1-4-25-21(24)19(14-16-8-6-5-7-9-16)22-20(23)18-12-10-17(11-13-18)15(2)3/h5-9,15,17-19H,4,10-14H2,1-3H3,(H,22,23)/t17-,18-,19-/m1/s1 |
| SMILES | CCOC(=O)[C@@H](Cc1ccccc1)NC(=O)[C@@H]2CC[C@H](CC2)C(C)C |
| IUPAC Name | ethyl (2R)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?