missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-pyrimidinecarboxylate, 97%, Thermo Scientific™

Catalog Number p-3770739
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
40929-50-8
C7H8N2O2
152.15
MFCD03094043
YCWDQAKDVQNVAR-UHFFFAOYSA-N
ethyl 5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, ethyl ester, pyrimidine-5-carboxylic acid ethyl ester, ethyl5-pyrimidinecarboxylate, 5-ethoxycarbonylpyrimidine, ethylpyrimidine-5-carboxylate, 2-ethylpyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid ethyl ester
4223810
ethyl pyrimidine-5-carboxylate
CCOC(=O)C1=CN=CN=C1
This item is not returnable. View return policy
40929-50-8
C7H8N2O2
152.15
MFCD03094043
YCWDQAKDVQNVAR-UHFFFAOYSA-N
ethyl 5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, ethyl ester, pyrimidine-5-carboxylic acid ethyl ester, ethyl5-pyrimidinecarboxylate, 5-ethoxycarbonylpyrimidine, ethylpyrimidine-5-carboxylate, 2-ethylpyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid ethyl ester
4223810
ethyl pyrimidine-5-carboxylate
CCOC(=O)C1=CN=CN=C1
Density 1.1600g/mL
Boiling Point 102°C (12.0 mmHg)
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.16
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Ethyl 5-pyrimidinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 5-pyrimidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'