missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebis(isothiouronium bromide), 98%, Thermo Scientific™

Product Code. p-7044498
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11316996

Brand: Thermo Scientific Alfa Aesar L06784.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6943-65-3
Molecular Formula C4H12Br2N4S2
Molecular Weight (g/mol) 340.096
MDL Number MFCD00035055
InChI Key VABTXEYLYWGJRB-UHFFFAOYSA-N
Synonym ethylenediisothiouronium dibromide, ethylene diisothiouronium dibromide, ethylenediisothiourea dihydrobromide, ethylene diisothiourea dihydrobromide, pseudourea, 2,2-ethylenedithiodi-, dihydrobromide, 2,2'-ethylenebisisothiouronium dibromide, carbamimidothioic acid, 1,2-ethanediyl ester, dihydrobromide, ethylenebis isothiouronium bromide, 2,2'-ethylene-bis-2-thiopseudourea , dihydrobromide, pseudourea, 2,2'-ethylenebis 2-thio-, dihydrobromide
PubChem CID 81370
IUPAC Name 2-carbamimidoylsulfanylethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)SC(=N)N.Br.Br

Specifications

Melting Point 224°C to 226°C
Quantity 5 g
Beilstein 3914616
Sensitivity Hygroscopic
Formula Weight 340.09
Percent Purity 98%
Chemical Name or Material Ethylenebis(isothiouronium bromide)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.