missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution volumetric, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Quantity | 500 mL |
Linear Formula | [-CH2N(CH2CO2Na)CH2CO2H]2 |
UN Number | NONH for all modes of transport |
Beilstein | 3822669 |
IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate |
Formula Weight | 336.21g/mol |
Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'