missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10335410
Click to view available options
Quantity:
250 g
500 g
This item is not returnable. View return policy
60-00-4
C10H16N2O8
292.24
3541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
This item is not returnable. View return policy
60-00-4
C10H16N2O8
292.24
3541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Melting Point 220°C
Color White
pH 2.5
Physical Form Powder/Solid
Quantity 250 g
Formula Weight 292.23g/mol
Packaging Plastic powder jar
Vapor Pressure 0.013 hPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'