Learn More
IKA™ EUROSTAR 40 Digital Overhead Stirrer
GREENER_CHOICE
Product Code. 13440567 Shop All IKA Products
Change view
Click to view available options
Plug Type:
EU
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
13440567 EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 13440567 Supplier IKA™ Supplier No. 0004444000
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Universal laboratory stirrer designed for simple stirring tasks for quantities up to 25 L (H2O).
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
TRUSTED_SUSTAINABILITY
Specifications
Speed 0/30 to 2000 rpm
Capacity 25 L
Display Digital
Voltage 100/115/230 V
Depth (Metric) Interior 208 mm
Diameter (Metric) Shaft 11 mm
Height (Metric) 248 mm
Top Plate Diameter (Metric) 16 mm
Width (Metric) 86 mm
Frequency 50/60 Hz
Plug Type EU
Certifications/Compliance DIN EN 60529
Material Alu-Cast Coating/Thermoplastic Polymer
Model Eurostar 40 digital
Protection Class IP40
Torque 40 Ncm
Type Stirrer
Viscosity 30,000 mPas
Weight (Metric) 4.4 kg