Learn More
Everolimus, Thermo Scientific™
A rapamycin analog
Brand:  Thermo Scientific Alfa Aesar J60139.MA
Quantity 10mg
 View more versions of this product
Product Code. 15482959
  • ONLINE EXCLUSIVE PRICE
    248.00€
    223.65€ / 10mg
    Save 24.35€
    9% Off
 Restricted Item
In Stock
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in dimethysulfoxide,ethanol and chloroform. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Identifiers
159351-69-6
958.24
HKVAMNSJSFKALM-PWMUVMERSA-N
CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC
C53H83NO14
MFCD07785165
129317877
Specifications
159351-69-6
MFCD07785165
14,3907
HKVAMNSJSFKALM-PWMUVMERSA-N
958.24
958.22
C53H83NO14
10mg
Soluble in dimethysulfoxide,ethanol and chloroform; Slightly soluble in water.
CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC
129317877
Everolimus
Product Suggestions
Product Suggestions
Videos
SDS Documents