Learn More
EVOS™ EVOS™ Condenser Slider, Meniscus A (clear liquids)
Even out field of view
Brand: EVOS™ AMEPMN3
Additional Details : Weight : 2.00000kg
Description
- Used to improve illumination under certain conditions
- Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
- Meniscus A slider is intended for brightfield applications at 2x magnification with low-volume samples in multi-well plates and with clear liquids
- Meniscus B slider is intended for brightfield applications at 2x magnification with high-volume samples in multi-well plates and with semi-opaque liquids
Specifications
Meniscus A Condenser Slider | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
1 Ea. |
EVOS | |
Diffuser Slider, Pinhole Slider, Meniscus Sliders |
For Research Use Only. Not for use in diagnostic procedures.