Learn More
Invitrogen™ EVOS™ Light Cube, CFP
Product Code. 11947285
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. 11947285
Brand: Invitrogen™ AMEP4653
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The revolutionary design of the EVOS™ light cubes combines a high intensity, long lifetime light source (LED), condensing optics, dichroic mirror and filters into a compact, adjustable, easy-to-install accessory for fluorescence detection on the EVOS™ imaging systems and the Countess II FL.
  • Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
TRUSTED_SUSTAINABILITY
Specifications
Type CFP Fluorescent Light Cube
Product Line EVOS
For Use With (Equipment) Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes LED, Condensing Optics, Hard-coated Filters
Wavelength Ex: 445/45 Em: 510/42
Quantity 1 each