Learn More
EVOS™ EVOS™ 20X Objective, fluorite, LWD
Components manufactured to ensure publication-quality images
Brand:  EVOS™ AMEP4624
Additional Details : Weight : 0.01000kg
Product Code. 11937275
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
Specifications
EVOS LWD Fluorite Objective
EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Plan Fluorite
0.45
EVOS
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
20X
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.