Learn More
EVOS™ Olympus 60X Oil Objective, apochromat, coverslip-corrected
Click to view available options
:
Plan Apochromat CC Objective Lens
Description
- Can be used to view images with different magnification
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Specifications
| Type | Objective Lens |
| Product Line | Olympus |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates |
| Lens Type | Plan Apochromat |
| Magnification | 60X |
| Quantity | 1 each |