Learn More
EVOS™ EVOS™ Onstage Vessel Holder, one 60-mm Petri dish
Allows user to utilize different types of vessels for imaging
Brand: EVOS™ AMEPVH031
Additional Details : Weight : 1.00000kg
Description
- Maximizes choice of cell culture vessels when conducting time-lapse imaging experiments using EVOS FL Auto Imaging System equipped with EVOS Onstage Incubator
- Allows perfect fit of micro-well plates or cell culture flasks/dishes to master plate of EVOS Onstage Incubator
Specifications
Vessel Holder | |
EVOS™ FL Auto Imaging System | |
1 60 mm Petri Dish |
EVOS | |
one, 60 mm Petri dish | |
1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.