Learn More
EVOS™ EVOS™Vessel Holder, Greiner/BD T-25 flask
Click to view available options
Quantity:
1 each
Unit Size:
Each
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
Specifications
Specifications
| Type | Vessel Holder |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Format | Greiner/BD T-25 flask |
| Holds | 1 25 cm2 BD/Greiner T-25 flask |
| Quantity | 1 each |