Learn More
Fisherbrand™ Excursion Trac Thermometer
Use this thermometer for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators. Fisherbrand™ Traceable™ Excursion Trac Thermometers feature user-defined time intervals.
Brand:  Fisherbrand™
Additional Details : Weight : 0.39600kg
Product Code. 15254026
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
  • Stores up to 10 unique alarm events
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
  • Status indicators - Low battery, memory full, USB data transfer and active alarm state