Learn More
VACUUBRAND™ Exhaust Vapor Condenser Upgrade Kit For NT Series Pumps
Product Code. 12664287
Click to view available options
For Use With:
ME 16C NT, MD 12C NT, MV 10C NT
Unit Size:
1 set
Product Code. 12664287
Brand: VACUUBRAND™ 20699948
Only null left
Specifications
For Use With ME 16C NT, MD 12C NT, MV 10C NT
Quantity 1 Kit
Type Exhaust Vapor Condenser Upgrade Kit