missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC6 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30060.100uL
Additional Details : Weight : 0.02000kg
Description
Rabbit polyclonal antibody raised against synthetic peptide of human EXOSC6.
- High specificity
- High reactivity
Specifications
EXOSC6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of human EXOSC6. | |
In PBS (2% sucrose, 0.09% sodium azide). | |
EAP4/MTR3/Mtr3p/hMtr3p/p11 | |
Rabbit | |
Protein A purification | |
RUO | |
118460 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
NM_058219 | |
EXOSC6 | |
A synthetic peptide corresponding to N-terminus of human EXOSC6. | |
100 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only (RUO)