Learn More
Sartorius External Calibration with HTML Evaluation
Shop All Sartorius Products
Click to view available options
:
External Calibration with HTML Evaluation
Description
- Features:
- Workflow has two levels:
- Administrator and User
- Administrators are allowed to define up to 5 weight values, including their ID, value and tolerances
- Users allowed to start calibration process
- Administrator and User
- Data acquisition and evaluation done automatically
- Documentation can be performed on a Sartorius printer and/or via Ethernet on basis of Q-Web via HTML visualization
- Workflow has two levels:
Specifications
Specifications
| For Use With (Equipment) | Cubis Balances |
| Type | Calibration Software |