missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific™ EZ-Link TFPA-PEG3-Biotin is an efficient, photoactivatable reagent based on tetrafluorophenyl azide for biotinylation, and which includes a 3-unit polyethylene glycol (PEG) spacer arm.
Highlights:
- Ideal photo-reactive biotin-labeling agent containing the efficient tetraflurophenyl azide (TFPA) moiety
- Fluorinated aryl azide inserts into active carbon-hydrogen bonds (C-H) or unsaturated carbon chains with greater efficiency than non-fluorinated aryl azides
- Hydrophilic polyethylene glycol (PEG) spacer contributes to improved solubility of the labeled protein
- Flexible, non-immunogenic PEG spacer imparts a reduced tendency of the labeled protein or peptide to aggregate in solution
- Photoactivation is achieved with a UV lamp that irradiates in the range of 300-370nm (320nm optimum)
Specifications
Specifications
| Product Type | TFPA-PEG3-Biotin |
| Quantity | 25 mg |
| Content And Storage | Store desiccated at 4°C. Shipped at ambient temperature. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Permeant |
| Chemical Reactivity | Light-activated |
| Reactive Moiety | Aryl Azide |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) |
| Label or Dye | Biotin |
| Spacer | Extra-long, Pegylated |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?