missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab')2-Goat anti-Human IgG, IgM (H+L) Secondary Antibody, Functional Grade, eBioscience™

F(ab')2-Goat Polyclonal Secondary Antibody

372.00€ - 3305.00€

Specifications

Antigen Human IgG, IgM (H+L)
Concentration 1 mg/mL
Content And Storage 4°C
Applications Flow Cytometry, Functional Assay
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17191262
View Documents
Invitrogen™
16-5099-38
5 mg
3305.00€
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15587786
View Documents
Invitrogen™
16-5099-85
500 μg
372.00€
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This polyclonal F(ab')2 fragment recognizes human IgM and IgG. Reactivity to bovine has been removed. This polyclonal antibody can be used to crosslink and activate B lymphocytes. Applications Reported: This polyclonal F(ab')2 Goat Anti-Human IgM+IgG Functinoal Grade Purified has been reported for use in functional assays (such as B cell activation). Concentrations should be determined empirically for optimal performance in the assay of interest. Applications Tested: F(ab')2 Anti-Human IgM+IgG Functional Grade Purified has been tested by in vitro stimulation of normal human peripheral blood cells. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Human IgG, IgM (H+L)
4°C
Polyclonal
Liquid
IgG
Human
Secondary
Affinity chromatography
1 mg/mL
Flow Cytometry, Functional Assay
Functional Grade
Goat
RUO
PBS with no preservative; pH 7.2
Secondary Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.