Learn More
Invitrogen™ F(ab')2-Goat anti-Rat IgG Fc Secondary Antibody, AP
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA129930
This item is not returnable.
View return policy
Description
PA1-29930 detects Rat IgG Fc from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rat IgG Fc | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| IgG fraction | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG F(ab')2 |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| TBS with 1% BSA, 1mM MgCl2, 50% glycerol, 0.1mM ZnCl2 and 0.05% sodium azide; pH 8 | |
| Rat IgG Fc fragment. | |
| 250 μg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |