missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAAH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62723
385.60 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
FAAH2 Polyclonal antibody specifically detects FAAH2 in Human samples. It is validated for Western Blot
Specifications
FAAH2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
AMDD, amidase domain containing, Amidase domain-containing protein, Anandamide amidohydrolase 2, EC 3.5.1.99, FAAH-2, fatty acid amide hydrolase 2, fatty-acid amide hydrolase 2, FLJ31204, Oleamide hydrolase 2, RP11-479E16.1 | |
Synthetic peptides corresponding to FAAH2(fatty acid amide hydrolase 2) The peptide sequence was selected form the C terminal of FAAH2. Peptide sequence SPLWELIKWCLGLSVYTIPSIGLALLEEKLRYSNEKYQKFKAVEESLRKE. The peptide sequence for this immunogen was taken from within the described region. | |
100 μg | |
metabolism | |
158584 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 1.0 μg/mL | |
Q6GMR7 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction