Learn More
Sino Biological™ FABP2 Recombinant Human Protein, His Tag
Expressed from DNA sequence encoding human FABP2 (Met 1-Asp 132) fused to a polyhistidine tag at C-terminus
Marque: Sino Biological™ 10130H08E5
Informations supplémentaires : Poids : 0.01000kg
Description
- Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Spécification
Lyophilized | |
16.6kd | |
5μg | |
−20°C | |
Fatty Acid Binding Protein 2, intestinal | |
Assay Standard | |
E.coli | |
His Tag |
2169 | |
≥95% by SDS-PAGE | |
Human | |
<1 EU/μg | |
FABP2 | |
Other proteins | |
FABP2 Recombinant Human Protein |
For Research Use Only. Not for use in diagnostic procedures.