Learn More
Fisherbrand™ FB65464 Rotary Vane Vacuum Pump
Two-stage rotary vane vacuum pump. Fisherbrand™ FB65464 Rotary Vane Vacuum Pump designed for laboratory applications requiring a deep vacuum.
Brand:  Fisherbrand™
Additional Details : Weight : 35.00000kg
Product Code. 12951141
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
These two stage rotary vane pumps provide a high water vapour tolerance. The low weight, small footprint and high pumping speed, together with increased chemical resistance, are ideal features for both research and routine applications.
  • Compact design, low weight and easy to use
  • Quiet running and excellent ultimate vacuum
  • Active corrosion protection
  • Easy maintenance
  • Supplied with clamps and seals for the inlet and outlet
  • Each pumps comes with anti-suck back valve, gas ballast valve, oil charging, centring ring, clamping ring and motor overload protection For a wide range of laboratory applications and continuous running.
Specifications
Specifications
2 x 10-3 mbar
115/230 V 50/60 Hz motor with Schuko UK and US power leads
170 mm
35 kg
260 mm
Rotary Vane Vacuum Pump
100/230 V
183/220 L/min.
54 x 17 x 26 cm (21 x 6.69 x 10.2 in.)
FB65464
50/60 Hz
540 mm
183 L/min.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products