missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FBXO33 293T Cell Transient Overexpression Lysate

Product Code. 18555161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
18555161 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 18555161

Brand: Novus Biologicals™ H00254170T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Cell Biology
Target FBXO33
Synonym BMND12, c14_5247, F-box only protein 33, F-box protein 33, Fbx33
Gene Symbol FBXO33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.