Learn More
Value Plastics Female Luer Thread Style Caps
Product Code. p-200105062 Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Clear Polycarbonate
White Nylon
Unit Size:
Pack of 25
Product Code. 30052968
Brand: Value Plastics 5012053
Only null left
Add to basket
Add to basket
Seal luer ports with a secure and leak-proof connection
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Thread design provides a secure closure—preventing contamination
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Packaging Quantity 25/Pk.
Inlet Female Luer Thread, Cap
Material (Seal) White Nylon