Learn More
Cole-Parmer FEP Tubing
Product Code. 30053278 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
7/16 in.
Diameter (English) Outer:
1/2 in.
1/4 in.
1/8 in.
3/16 in.
5/32 in.
9/16 in.
Unit Size:
15m
Product Code. 30053278
Brand: Cole-Parmer 5011924
Only null left
Add to basket
Add to basket
Chemically inert, thermal-resistant rigid tubing with optical clarity
Made of 100% virgin-grade fluorinated ethylene propylene (FEP). Excellent chemical and thermal resistance. Great optical clarity—easily view your sample in use. Sterilize by autoclave, EtO, and chemical sterilants. Complies with FDA 21 CFR 177.1550, USP Class VI, NSF 51, NSF 61, NSF 372, and ASTM D2116. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Certifications/Compliance ADCF;FDA;NSF-51;NSF-61;RoHS;USP
Diameter (English) Inner 1/2 in.
Diameter (English) Outer 9/16 in.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.