Learn More
BUCHI FEP Tubing
For use in preparative chromatography
Brand:  BUCHI 044346
Additional Details : Weight : 0.05000kg
Product Code. 11937258
  • 193.00€ / 1m
Estimated Shipment: 23-05-2024
to see stock.
Specifications
Tubing
3.18 mm
FEP
Natural
5 m