missing translation for 'onlineSavingsMsg'
Learn More

Human FER, His Tag Recombinant Protein

FER Recombinant Human Protein

Brand:  Invitrogen PV3806

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 11593537

  • 632.00€ / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FER is a member of the FPS⁄FES family of non-receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors.
Specifications

Specifications

NP_005237
2241
10 μg
FER Kinase Family
Human
GST Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic
Dry Ice
FER
Baculovirus
Recombinant, Catalytic Domain, Cytoplasmic
Cytoplasmic Tyrosine Kinases
Kinase Assay
60.3 kDa
FER, TYK3
aa 541-822
Recombinant
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton™ X-100, 2 mM DTT and 50% glycerol

Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Tyrosine Kinases
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
fer (fps⁄fes related) tyrosine kinase
Kinases & Inhibitors
Liquid
Signal Transduction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions