missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FER, His Tag Recombinant Protein
FER Recombinant Human Protein
632.00€
Specifications
Accession Number | NP_005237 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 60.3 kDa |
Gene Alias | FER, TYK3 |
KinaseFamily | FER Kinase Family |
Description
FER is a member of the FPS⁄FES family of non-receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors.Specifications
NP_005237 | |
60.3 kDa | |
FER Kinase Family | |
Recombinant | |
Dry Ice |
Kinase Assay | |
FER, TYK3 | |
Human | |
Tyrosine Kinases | |
Baculovirus |