missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EcoO109I (DraII) (10 U/μL)

Cut at RG^GNCCY sites with EcoO109I (DraII) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: DraII).

Brand:  Thermo Scientific™ ER0261

Additional Details : Weight : 0.00900kg

Product Code. 10242630

  • 88.75€ / 2000 units
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lambda DNA digested with EcoO109I (DraII), 0.7% agarose, 3 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
EcoO109I (DraII)
Yes
No
Traditional Cloning
Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
10X Buffer Tango
37°C
Restriction Enzyme
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.