Learn More
Thermo Scientific™ XmiI (AccI) (10 U/μL)
Product Code. p-4522204 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2,000 units
400 U
Unit Size:
2000 units
400 units
Product Code. 10706031
Brand: Thermo Scientific™ ER1481
Cut at GT^MKAC sites with XmiI (AccI) restriction enzyme, which functions best at 37°C in B buffer (Isoschizomers: AccI, FblI).
Lambda DNA digested with XmiI, 1% agarose, 9 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme XmiI (AccI)
Compatible Buffer 10X Buffer B
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 400 U
Product Type Restriction Enzyme
Research Category Traditional Cloning