missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Fetal Bovine Serum (FBS)

Product Code. 15799170 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15799170

Brand: MP Biomedicals 092910154

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Used in a broad range of cell culture applications for promoting/enhancing growth/survival.

  • FBS provides many non-defined growth promoting and survival enhancing factors to cells in culture.
  • For cell culture, mammalian cell culture, source of growth promoting and survival enhancing factor.
  • Golden brown liquid.
  • Sterile.
TRUSTED_SUSTAINABILITY

Specifications

Color Golden brown
Content And Storage Store at -20°C
Form Liquid
Quantity 500 mL
Concentration Protein: 3.4 - 4.6%
Sterility Sterile
Formulation Liquid; Glucose: 137 mg/dL; Sodium: 140 meq/L; Potassium: 10.6 meq/L; Chloride: 103 meq/L; B.U.N.: 15 mg/dL; Creatinine: 2.7 mg/dL; Uric Acid: 2.3 mg/dL; Calcium: 14.0 mg/dL; Phosphorus: 9.9 mg/dL; Albumin: 1.9 gm/dL; Total Bilirubin: 0.2 mg/dL; ALT (SGPT): 5 IU/L; Alkaline phosphatase: 261 IU/L; AST (SGOT): 43 IU/L; LDH: 556 IU/L; Cholesterol: 31 mg/dL; Triglycerides: 57 mg/dL.
For Use With (Application) Cell culture. Mammalian cell culture. Source of growth promoting and survival enhancing factor.
pH pH 6.7-7.6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.