Learn More
Coverguard™ FFP1 NR D Face Mask
Product Code. 12003186
Change view
Click to view available options
Style:
Flat Foldable Half-Mask
Flat Foldable Half-Mask with Valve
Shell Mask
Shell Mask with Valve
Nosepiece Type:
Adjustable Nose Clip
Adjustable Nose Clip with PU Foam Nose Pad
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 120
Pack of 240
Product Code. Style Nosepiece Type With Exhalation Valve unitSize
12003186 Flat Foldable Half-Mask Adjustable Nose Clip No Pack of 240
12013186 Shell Mask Adjustable Nose Clip with PU Foam Nose Pad No Pack of 240
12023186 Flat Foldable Half-Mask with Valve Adjustable Nose Clip Yes Pack of 240
12033186 Shell Mask with Valve Adjustable Nose Clip with PU Foam Nose Pad Yes Pack of 120
4 options
Product Code. 12003186
Brand: Coverguard™ 23100
Only null left
Add to basket
Add to basket
The Coverguard™ FFP1 masks are single-use and protect against non-toxic solid and liquid aerosols, in applications such as farming, silviculture and fishing, light industries, maintenance, infrastructure and building, DIY, as well as general use in the lab or field.
  • PPE Cat. III FFP1 NR D
  • Disposable
  • Electrostatic filter media
  • Sliding elastic band for better adjustment
  • Latex-free, non-woven polypropylene
  • Adjustable nose clip
Specifications
Certifications/Compliance (Europe) CE PPE Category III FFP1 NR D, EN 149:2001 + A1:2009
Description FFP1 Disposable Foldable Half-Mask, Sealed Outer Contour
Disposable Single-use
Material Non-woven Polypropylene
With Exhalation Valve No
Filter Efficiency FFP1
Latex Free Yes
Material Headstrap Synthetic Rubber Elastic (Latex Free)
Nosepiece Type Adjustable Nose Clip
Resistant To Non-Toxic Aerosols and Non-Toxic Dusts
Sterility Non-sterile
Style Flat Foldable Half-Mask